This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPARCL1 SIRNA Assays ›
Gene Symbol:
Sparcl1
Gene Name:
SPARC-like 1 (hevin)
Gene Aliases:
Ecm2, Sc1
Chromosome Location:
Chr.14: 6994261 - 7025309 on Build Rnor_6.0
UniGene ID:
Rn.3091
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS303624
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sparcl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012946.1 NP_037078.1 4 1119
GenBank mRNA BC061755.1 - 4 1120 5600008
U27562.1 - 4 1119

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0