This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYP19A1 SIRNA Assays ›
Gene Symbol:
Cyp19a1
Gene Name:
cytochrome P450, family 19, subfamily a, polypeptide 1
Gene Aliases:
Aromatase, Cyp19, Cyp19a, p450arom
Chromosome Location:
Chr.8: 58744849 - 58772408 on Build Rnor_6.0
UniGene ID:
Rn.21402
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS303092
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyp19a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017085.2 NP_058781.2 9 1817

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

luteinization
estrogen biosynthetic process
bile acid metabolic process
C21-steroid hormone metabolic process
androgen metabolic process
male gonad development
response to herbicide
response to metal ion
response to cesium ion
response to ionizing radiation
response to organic cyclic compound
biphenyl metabolic process
dibenzo-p-dioxin metabolic process
phthalate metabolic process
hippocampus development
hypothalamus development
melatonin metabolic process
adrenal gland development
granulocyte differentiation
response to estradiol
response to testosterone
progesterone metabolic process
response to drug
response to morphine
response to estrogen
cellular response to fibroblast growth factor stimulus
negative regulation of bone resorption
regulation of synapse structure or activity
oxidation-reduction process
bone development
prostate gland growth
cellular response to glucose stimulus
cellular response to gonadotropin stimulus
cellular response to follicle-stimulating hormone stimulus
cellular response to hydrostatic pressure
iron ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
heme binding
aromatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header