This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GIPR SIRNA Assays ›
Gene Symbol:
Gipr
Gene Name:
gastric inhibitory polypeptide receptor
Gene Aliases:
Gippr, RATGIPPR
Chromosome Location:
Chr.1: 80063049 - 80073223 on Build Rnor_6.0
UniGene ID:
Rn.9676
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302941
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gipr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012714.1 NP_036846.1 7 658
GenBank mRNA L19660.1 - 7 658

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
positive regulation of cytosolic calcium ion concentration
response to glucose
positive regulation of cAMP biosynthetic process
endocrine pancreas development
positive regulation of insulin secretion
gastric inhibitory peptide signaling pathway
response to axon injury
response to calcium ion
response to fatty acid
gastric inhibitory peptide receptor activity
peptide hormone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0