This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYP2C12 SIRNA Assays ›
Gene Symbol:
Cyp2c12
Gene Name:
cytochrome P450, family 2, subfamily c, polypeptide 12
Gene Aliases:
Cyp2c40, P450pb1, RATP4515B2, RATP4515B8
Chromosome Location:
Chr.1: 258709726 - 258766873 on Build Rnor_6.0
UniGene ID:
Rn.2586
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302918
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyp2c12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031572.1 NP_113760.1 9 1394
GenBank mRNA BC089790.1 - 9 1402 7371667
FQ209627.1 - 6 833
FQ219491.1 - 8 1269
J03786.1 - 9 1394

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

xenobiotic metabolic process
epoxygenase P450 pathway
exogenous drug catabolic process
oxidation-reduction process
monooxygenase activity
iron ion binding
arachidonic acid epoxygenase activity
steroid hydroxylase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
oxygen binding
heme binding
aromatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0