This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLA2G4A SIRNA Assays ›
Gene Symbol:
Pla2g4a
Gene Name:
phospholipase A2, group IVA (cytosolic, calcium-dependent)
Gene Aliases:
Pla2c, Pla2g4, cPLA2
Chromosome Location:
Chr.13: 67062252 - 67206688 on Build Rnor_6.0
UniGene ID:
Rn.10162
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302600
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pla2g4a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133551.2 NP_598235.2 14, 15 1748
GenBank mRNA BC070940.1 - 14, 15 1748 7099813
S77829.1 - 14, 15 1734
U38376.1 - 14, 15 1749

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008869, BC003816, AK004701, M72394, AK144750.

Gene Ontology Categories:

Function(s) Process(es)

ovulation from ovarian follicle
luteolysis
aging
positive regulation of cell proliferation
phospholipid catabolic process
response to heat
response to hormone
response to organic substance
response to lithium ion
response to organonitrogen compound
arachidonic acid metabolic process
positive regulation of bone mineralization
positive regulation of vesicle fusion
positive regulation of prostaglandin biosynthetic process
positive regulation of fever generation
response to lipopolysaccharide
response to vitamin D
regulation of cell proliferation
response to hydrogen peroxide
positive regulation of apoptotic process
surfactant homeostasis
icosanoid biosynthetic process
decidualization
arachidonic acid secretion
positive regulation of inflammatory response
response to glucocorticoid
response to calcium ion
response to methylmercury
cellular response to antibiotic
lysophospholipase activity
phospholipase A2 activity
calcium ion binding
calcium-dependent phospholipid binding
histone acetyltransferase binding
calcium-dependent phospholipase A2 activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header