This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FMO2 SIRNA Assays ›
Gene Symbol:
Fmo2
Gene Name:
flavin containing monooxygenase 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 80752552 - 80775338 on Build Rnor_6.0
UniGene ID:
Rn.3928
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302491
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fmo2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144737.2 NP_653338.2 8 1309
XM_008769604.1 XP_008767826.1 7 1573
XM_008769605.1 XP_008767827.1 8 1383
GenBank mRNA AF458414.1 - 8 1276
BC070904.1 - 8 1309 7100688

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

organic acid metabolic process
NADP metabolic process
xenobiotic metabolic process
toxin metabolic process
drug metabolic process
oxidation-reduction process
NADPH oxidation
oxygen metabolic process
monooxygenase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header