This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LGALS3BP SIRNA Assays ›
Gene Symbol:
Lgals3bp
Gene Name:
lectin, galactoside-binding, soluble, 3 binding protein
Gene Aliases:
CyCAP, MAC2BP, Ppicap, mama
Chromosome Location:
Chr.10: 107415334 - 107424672 on Build Rnor_6.0
UniGene ID:
Rn.3251
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302358
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lgals3bp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139096.1 NP_620796.1 2, 3 204
GenBank mRNA AF065438.1 - 2, 3 204
AY552591.1 - 1, 2 66
BC081724.1 - 2, 3 177 7123606

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
cell adhesion
scavenger receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0