This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IDH1 SIRNA Assays ›
Gene Symbol:
Idh1
Gene Name:
isocitrate dehydrogenase 1 (NADP+), soluble
Gene Aliases:
-
Chromosome Location:
Chr.9: 71882108 - 71911626 on Build Rnor_6.0
UniGene ID:
Rn.3561
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302243
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Idh1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031510.1 NP_113698.1 8 1258
XM_006245049.2 XP_006245111.1 9 1316
XM_008767076.1 XP_008765298.1 9 1348
GenBank mRNA FQ210018.1 - 1 90
FQ214242.1 - 9 1339
L35317.1 - 8 1258

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glyoxylate cycle
tricarboxylic acid cycle
isocitrate metabolic process
2-oxoglutarate metabolic process
glutathione metabolic process
response to oxidative stress
female gonad development
response to organic cyclic compound
response to steroid hormone
regulation of phospholipid catabolic process
regulation of phospholipid biosynthetic process
magnesium ion binding
isocitrate dehydrogenase (NADP+) activity
receptor binding
protein homodimerization activity
NADP binding
NAD binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0