This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACE SIRNA Assays ›
Gene Symbol:
Ace
Gene Name:
angiotensin I converting enzyme
Gene Aliases:
Dcp1, StsRR92
Chromosome Location:
Chr.10: 94170766 - 94213831 on Build Rnor_6.0
UniGene ID:
Rn.10149
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302103
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ace
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012544.1 NP_036676.1 16 2458
GenBank mRNA AF201331.1 - 16 2458
AF201332.1 - 16 2458
AF532783.1 - 16 2402
AF532784.1 - 16 2402
AF539425.1 - 835
BC085760.1 - 16 2458 7097136
L36664.1 - 3 262
U03708.1 - 16 2458
U03734.1 - 16 2458

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
kidney development
neutrophil mediated immunity
regulation of systemic arterial blood pressure by renin-angiotensin
positive regulation of systemic arterial blood pressure
proteolysis
peptide metabolic process
spermatogenesis
brain development
female pregnancy
aging
regulation of blood pressure
male gonad development
posttranscriptional regulation of gene expression
negative regulation of gene expression
bradykinin catabolic process
regulation of smooth muscle cell migration
sensory perception of pain
lung development
organ regeneration
response to nutrient levels
negative regulation of protein binding
positive regulation of protein binding
response to lipopolysaccharide
response to laminar fluid shear stress
vasoconstriction
hormone catabolic process
response to drug
eating behavior
positive regulation of apoptotic process
peptide catabolic process
lung alveolus development
beta-amyloid metabolic process
arachidonic acid secretion
positive regulation of inflammatory response
positive regulation of neurogenesis
heart contraction
regulation of angiotensin metabolic process
angiogenesis involved in coronary vascular morphogenesis
positive regulation of protein tyrosine kinase activity
cellular response to glucose stimulus
response to dexamethasone
cell proliferation in bone marrow
negative regulation of calcium ion import
response to thyroid hormone
positive regulation of peptidyl-tyrosine autophosphorylation
regulation of hematopoietic stem cell proliferation
negative regulation of gap junction assembly
positive regulation of peptidyl-cysteine S-nitrosylation
actin binding
endopeptidase activity
carboxypeptidase activity
drug binding
peptidase activity
metallopeptidase activity
exopeptidase activity
tripeptidyl-peptidase activity
peptidyl-dipeptidase activity
zinc ion binding
chloride ion binding
mitogen-activated protein kinase kinase binding
bradykinin receptor binding
metal ion binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0