This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAMK2B SIRNA Assays ›
Gene Symbol:
Camk2b
Gene Name:
calcium/calmodulin-dependent protein kinase II beta
Gene Aliases:
Ck2b
Chromosome Location:
Chr.14: 86208876 - 86297751 on Build Rnor_6.0
UniGene ID:
Rn.9743
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS302053
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Camk2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042354.1 NP_001035813.1 5 381
NM_001042356.1 NP_001035815.1 5 381
NM_021739.2 NP_068507.2 5 381
XM_006251170.2 XP_006251232.1 5 480
XM_006251171.2 XP_006251233.1 5 480
XM_006251172.2 XP_006251234.1 5 480
XM_006251174.2 XP_006251236.1 5 480
XM_006251176.2 XP_006251238.1 5 461
XM_006251177.2 XP_006251239.1 5 460
GenBank mRNA CB616255.1 - 5 381
M16112.1 - 5 361
X83375.1 - 5 343

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_172081, NM_172078, NM_172082, NM_172083, NM_172084, and others.

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
inhibitory G-protein coupled receptor phosphorylation
protein phosphorylation
calcium ion transport
positive regulation of neuron projection development
peptidyl-serine phosphorylation
regulation of synaptic transmission, cholinergic
positive regulation of phospholipase A2 activity
response to cadmium ion
protein autophosphorylation
regulation of long-term neuronal synaptic plasticity
cell projection morphogenesis
neuromuscular process controlling balance
long-term synaptic potentiation
activation of meiosis involved in egg activation
positive regulation of dendritic spine morphogenesis
positive regulation of synapse maturation
positive regulation of apoptotic signaling pathway
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
protein homodimerization activity
phospholipase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0