This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APC SIRNA Assays ›
Gene Symbol:
Apc
Gene Name:
adenomatous polyposis coli
Gene Aliases:
RATAPC
Chromosome Location:
Chr.18: 27011706 - 27106323 on Build Rnor_6.0
UniGene ID:
Rn.156346
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301999
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Apc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012499.1 NP_036631.1 15 2027
XM_006254506.2 XP_006254568.1 16 2287
XM_006254508.2 XP_006254570.1 17 2128
XM_006254509.2 XP_006254571.1 17 2097
XM_006254510.2 XP_006254572.1 15 2233
XM_006254511.2 XP_006254573.1 16 2074
XM_006254512.2 XP_006254574.1 16 2736
XM_006254513.2 XP_006254575.1 15 2182
XM_006254515.2 XP_006254577.1 17 1953
XM_006254518.2 XP_006254580.1 16 2551
XM_006254520.2 XP_006254582.1 16 1984
XM_006254521.2 XP_006254583.1 16 1771
XM_008772015.1 XP_008770237.1 16 4219
XM_008772016.1 XP_008770238.1 16 4169
XM_008772017.1 XP_008770239.1 16 2560
GenBank mRNA D38629.1 - 15 2027

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
kidney development
hair follicle development
protein complex assembly
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
cell cycle arrest
metaphase/anaphase transition of mitotic cell cycle
mitotic spindle assembly checkpoint
establishment or maintenance of cell polarity
pattern specification process
axonogenesis
negative regulation of cell proliferation
axis specification
anterior/posterior pattern specification
dorsal/ventral pattern formation
proximal/distal pattern formation
Wnt signaling pathway
cell migration
stem cell population maintenance
negative regulation of Wnt signaling pathway
regulation of cell migration
positive regulation of cell migration
regulation of epithelial cell differentiation
positive regulation of epithelial cell differentiation
pancreas development
positive regulation of microtubule polymerization
cytoplasmic microtubule organization
neuron projection development
positive regulation of pseudopodium assembly
regulation of microtubule-based process
T cell differentiation in thymus
somatic stem cell population maintenance
negative regulation of odontogenesis
response to drug
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of MAPK cascade
skin development
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
regulation of cell differentiation
positive regulation of cell differentiation
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cell adhesion
muscle cell cellular homeostasis
thymus development
negative regulation of epithelial cell proliferation
regulation of nitrogen compound metabolic process
chromosome organization
regulation of cell cycle
positive regulation of cell division
regulation of attachment of spindle microtubules to kinetochore
retina development in camera-type eye
canonical Wnt signaling pathway
negative regulation of epithelial cell proliferation involved in prostate gland development
negative regulation of canonical Wnt signaling pathway
response to alcohol
cellular response to nerve growth factor stimulus
regulation of glutamate metabolic process
protease binding
protein binding
beta-catenin binding
microtubule binding
protein kinase regulator activity
protein kinase binding
protein complex binding
gamma-catenin binding
cadherin binding
microtubule plus-end binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0