This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDK12 SIRNA Assays ›
Gene Symbol:
Cdk12
Gene Name:
cyclin-dependent kinase 12
Gene Aliases:
Crk7, Crkrs, Pksc
Chromosome Location:
Chr.10: 86157188 - 86222535 on Build Rnor_6.0
UniGene ID:
Rn.48817
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301905
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdk12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033867.1 NP_001029039.1 1 822
NM_138916.2 NP_620271.1 1 822
XM_006247250.2 XP_006247312.1 1 1376
XM_006247251.2 XP_006247313.1 1 1376
XM_006247253.2 XP_006247315.1 1 1375
GenBank mRNA AY072294.1 - 1 822
AY962568.1 - 1 822

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
regulation of MAP kinase activity
regulation of RNA splicing
protein autophosphorylation
regulation of cell cycle
phosphorylation of RNA polymerase II C-terminal domain
protein kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity
cyclin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0