This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRSS8 SIRNA Assays ›
Gene Symbol:
Prss8
Gene Name:
protease, serine, 8
Gene Aliases:
CAP1
Chromosome Location:
Chr.1: 199372519 - 199377035 on Build Rnor_6.0
UniGene ID:
Rn.107364
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301784
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prss8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138836.1 NP_620191.1 6 1034
GenBank mRNA AB017638.1 - 6 1034
AF202076.1 - 6 1032
BC061800.1 - 6 1047 5598332

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hair follicle development
proteolysis
positive regulation of sodium ion transport
response to drug
response to peptide hormone
response to mineralocorticoid
transepithelial transport
serine-type endopeptidase activity
serine-type peptidase activity
sodium channel regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0