This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PACS1 SIRNA Assays ›
Gene Symbol:
Pacs1
Gene Name:
phosphofurin acidic cluster sorting protein 1
Gene Aliases:
Pacs-1
Chromosome Location:
Chr.1: 220515117 - 220644825 on Build Rnor_6.0
UniGene ID:
Rn.154425
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301671
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pacs1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134406.1 NP_599233.1 10, 11 1390
XM_006230657.2 XP_006230719.1 10, 11 1479
GenBank mRNA AF076183.1 - 10, 11 1390
AF076184.1 - 10, 11 1390

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to Golgi
positive regulation of protein binding
cellular protein localization
protein targeting to plasma membrane
ion channel binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0