This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AGPAT4 SIRNA Assays ›
Gene Symbol:
Agpat4
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 48719632 - 48826302 on Build Rnor_6.0
UniGene ID:
Rn.41595
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301320
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Agpat4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133406.1 NP_596897.1 6 937
XM_006227856.2 XP_006227918.1 6 1652
XM_006227857.2 XP_006227919.1 6 1306
GenBank mRNA AB067572.1 - 6 937
BC086992.1 - 6 856 7191675

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026644, AK166917, BC047281, AK005139.

Gene Ontology Categories:

Function(s) Process(es)

CDP-diacylglycerol biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0