This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNRK SIRNA Assays ›
Gene Symbol:
Snrk
Gene Name:
SNF related kinase
Gene Aliases:
-
Chromosome Location:
Chr.8: 130749857 - 130805225 on Build Rnor_6.0
UniGene ID:
Rn.10245
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301264
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snrk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138833.1 NP_620188.1 6 1269
XM_006244057.2 XP_006244119.1 7 1315
XM_006244058.2 XP_006244120.1 7 1349
XM_006244059.1 XP_006244121.1 7 1308
XM_006244060.2 XP_006244122.1 6 1323
XM_006244061.1 XP_006244123.1 6 1329
XM_006244062.2 XP_006244124.1 6 1257
XM_008766666.1 XP_008764888.1 6 1248
XM_008766667.1 XP_008764889.1 6 1390
GenBank mRNA X89383.1 - 6 1269

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001164572, NM_133741, AF387809, BC150742, BC020189, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of neuron apoptotic process
magnesium ion binding
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0