This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANP32B SIRNA Assays ›
Gene Symbol:
Anp32b
Gene Name:
acidic (leucine-rich) nuclear phosphoprotein 32 family, member B
Gene Aliases:
PAL31
Chromosome Location:
Chr.5: 62070709 - 62094633 on Build Rnor_6.0
UniGene ID:
Rn.20465
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301244
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Anp32b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131911.2 NP_571986.1 3 446
XM_008763695.1 XP_008761917.1 4 758
GenBank mRNA AB025581.1 - 3 283
BC086508.1 - 3 446 7103152

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_130889, AB025582, BC093506, AK136648, BC005628, and others.

Gene Ontology Categories:

Function(s) Process(es)

vasculature development
nucleosome assembly
activation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of cell proliferation
ventricular system development
negative regulation of cell differentiation
positive regulation of protein export from nucleus
inner ear development
palate development
positive regulation of G1/S transition of mitotic cell cycle
histone binding
RNA polymerase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0