This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLCD4 SIRNA Assays ›
Gene Symbol:
Plcd4
Gene Name:
phospholipase C, delta 4
Gene Aliases:
-
Chromosome Location:
Chr.9: 81814772 - 81858397 on Build Rnor_6.0
UniGene ID:
Rn.37434
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS301037
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Plcd4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080688.1 NP_542419.1 9 1384
XM_006245135.2 XP_006245197.1 10 2459
XM_006245136.2 XP_006245198.1 9 1300
XM_006245137.2 XP_006245199.1 9 1599
XM_006245138.2 XP_006245200.1 9 1474
XM_008767182.1 XP_008765404.1 9 1878
XM_008767183.1 XP_008765405.1 9 1878
XM_008767184.1 XP_008765406.1 9 1917
XM_008767185.1 XP_008765407.1 9 1301
XM_008767186.1 XP_008765408.1 9 1879
XM_008767187.1 XP_008765409.1 9 1879
XM_008767188.1 XP_008765410.1 9 1878
XM_008767189.1 XP_008765411.1 9 1878
XM_008767190.1 XP_008765412.1 9 1877
XM_008767191.1 XP_008765413.1 9 1877
GenBank mRNA U16655.1 - 9 1384

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

acrosome reaction
lipid catabolic process
intracellular signal transduction
phosphatidylinositol metabolic process
phosphatidylinositol phospholipase C activity
signal transducer activity
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0