This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BAIAP2 SIRNA Assays ›
Gene Symbol:
Baiap2 LOC102555951
Gene Name:
BAI1-associated protein 2
brain-specific angiogenesis inhibitor 1-associated protein 2-like
Gene Aliases:
Irsp53
-
Chromosome Location:
Chr.10: 109107326 - 109187458 on Build Rnor_6.0
Chromosome Location:
Chr.10: 109161505 - 109163625 on Build Rnor_6.0
UniGene ID:
Rn.95155
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS300919
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Baiap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057196.1 NP_476544.1 9 950
XM_006247869.2 XP_006247931.1 9 1013
XM_006247870.2 XP_006247932.1 9 1013
XM_008768464.1 XP_008766686.1 9 1013
GenBank mRNA AY037934.1 - 9 950
BC074009.1 - 9 984 7099884
Gene Symbol: LOC102555951
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247941.2 XP_006248003.1 2 157

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001037755, NM_130862, AK049469, BC006620, AF390178, and others.

Gene Ontology Categories:

Function(s) Process(es)

plasma membrane organization
regulation of cell shape
response to bacterium
dendrite development
regulation of actin cytoskeleton organization
regulation of synaptic plasticity
actin filament bundle assembly
actin crosslink formation
positive regulation of dendritic spine morphogenesis
protein binding
cytoskeletal adaptor activity
PDZ domain binding
identical protein binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0