This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCNF SIRNA Assays ›
Gene Symbol:
Ccnf
Gene Name:
cyclin F
Gene Aliases:
-
Chromosome Location:
Chr.10: 13594687 - 13619935 on Build Rnor_6.0
UniGene ID:
Rn.15126
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300899
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccnf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100474.1 NP_001093944.1 14, 15 1622
GenBank mRNA AF410816.1 - 14, 15 1622

Back To Top

More Information


Gene Ontology Categories:

Function(s)

re-entry into mitotic cell cycle
placenta development
mitotic nuclear division
negative regulation of centrosome duplication
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
cell division

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0