This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMC1 SIRNA Assays ›
Gene Symbol:
Psmc1
Gene Name:
proteasome (prosome, macropain) 26S subunit, ATPase, 1
Gene Aliases:
s4
Chromosome Location:
Chr.6: 124123283 - 124135644 on Build Rnor_6.0
UniGene ID:
Rn.10526
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300843
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057123.1 NP_476464.1 9 1135
GenBank mRNA BC063157.1 - 10 1147 6919865
D50696.1 - 9 1135
FQ209916.1 - 10 1140

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_002802, AK299121, L02426, AB451441, AK222668, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein catabolic process
ATP binding
ATPase activity
TBP-class protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0