This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTPN2 SIRNA Assays ›
Gene Symbol:
Ptpn2
Gene Name:
protein tyrosine phosphatase, non-receptor type 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 63415298 - 63489266 on Build Rnor_6.0
UniGene ID:
Rn.33497
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300748
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptpn2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053990.1 NP_446442.1 7 749
XM_006254854.2 XP_006254916.1 7 1988
XM_006254855.2 XP_006254917.1 7 1988
XM_008772080.1 XP_008770302.1 7 1988
XM_008772081.1 XP_008770303.1 7 1988
GenBank mRNA BC078758.1 - 7 788 7132485
X58828.1 - 7 749

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
negative regulation of cell proliferation
insulin receptor signaling pathway
negative regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of lipid storage
B cell differentiation
T cell differentiation
erythrocyte differentiation
peptidyl-tyrosine dephosphorylation
negative regulation of epidermal growth factor receptor signaling pathway
negative regulation of tyrosine phosphorylation of Stat1 protein
negative regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of tyrosine phosphorylation of Stat5 protein
negative regulation of tyrosine phosphorylation of Stat6 protein
glucose homeostasis
negative regulation of macrophage differentiation
positive regulation of gluconeogenesis
negative regulation of insulin receptor signaling pathway
negative regulation of inflammatory response
negative regulation of T cell receptor signaling pathway
negative regulation of chemotaxis
negative regulation of interferon-gamma-mediated signaling pathway
negative regulation of type I interferon-mediated signaling pathway
negative regulation of interleukin-6-mediated signaling pathway
negative regulation of ERK1 and ERK2 cascade
regulation of hepatocyte growth factor receptor signaling pathway
negative regulation of interleukin-2-mediated signaling pathway
negative regulation of prolactin signaling pathway
negative regulation of interleukin-4-mediated signaling pathway
negative regulation of macrophage colony-stimulating factor signaling pathway
negative regulation of positive thymic T cell selection
negative regulation of platelet-derived growth factor receptor-beta signaling pathway
DNA binding
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
integrin binding
protein kinase binding
syntaxin binding
receptor tyrosine kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header