This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPS1 SIRNA Assays ›
Gene Symbol:
Gps1
Gene Name:
G protein pathway suppressor 1
Gene Aliases:
MFH
Chromosome Location:
Chr.10: 109966875 - 109971912 on Build Rnor_6.0
UniGene ID:
Rn.16873
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300713
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gps1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270939.1 NP_001257868.1 2 86
NM_053969.3 NP_446421.3 2 260
XM_006247865.1 XP_006247927.1 2 255
XM_006247866.1 XP_006247928.1 2 255
XM_006247867.2 XP_006247929.1 2 115
XM_008768462.1 XP_008766684.1 2 255
XM_008768463.1 XP_008766685.1 2 115
GenBank mRNA BC061746.1 - 2 209 5599712
CB771566.1 - 2 260
CB792560.1 - 2 136
X87885.1 - 1 41

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cullin deneddylation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0