This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RIMS2 SIRNA Assays ›
Gene Symbol:
Rims2
Gene Name:
regulating synaptic membrane exocytosis 2
Gene Aliases:
Nim2, Rim2
Chromosome Location:
Chr.7: 78092020 - 78594162 on Build Rnor_6.0
UniGene ID:
Rn.161948
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300662
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rims2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053945.2 NP_446397.1 25, 26 4141
NM_145881.2 NP_665888.1 2, 3 463
GenBank mRNA AF199322.1 - 25, 26 4141
AF199323.1 - 20, 21 3532
AF199324.1 - 20, 21 3673
AF199325.1 - 21, 22 3580
AF199326.1 - 20, 21 3532
AF199327.1 - 20, 21 3532
AF199328.1 - 19, 20 3352
AF199329.1 - 19, 20 3466
AF199330.1 - 24, 25 4093
AF199331.1 - 23, 24 4051
AF199332.1 - 20, 21 3532
AF199335.1 - 2, 3 464
AF548738.1 - 23, 24 3564

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
exocytosis
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of inhibitory postsynaptic potential
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
protein domain specific binding
ion channel binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0