This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other INPP4B SIRNA Assays ›
Gene Symbol:
Inpp4b
Gene Name:
inositol polyphosphate-4-phosphatase, type II
Gene Aliases:
-
Chromosome Location:
Chr.19: 29592889 - 30341442 on Build Rnor_6.0
UniGene ID:
Rn.10829
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300603
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Inpp4b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053917.1 NP_446369.1 18 2184
XM_008772462.1 XP_008770684.1 18 2027
XM_008772463.1 XP_008770685.1 18 2027
XM_008772464.1 XP_008770686.1 18 2184
XM_008772465.1 XP_008770687.1 17 1976
GenBank mRNA U96920.1 - 18 2184
U96921.1 - 18 2184

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
biological_process
dephosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
negative regulation of osteoclast differentiation
regulation of nucleocytoplasmic transport
regulation of bone remodeling
regulation of protein kinase B signaling
lipid binding
phosphatidylinositol-3,4-bisphosphate 4-phosphatase activity
phosphatidylinositol bisphosphate phosphatase activity
phosphatidylinositol trisphosphate phosphatase activity
phosphatidylinositol-4,5-bisphosphate 4-phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0