This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DYNC1I2 SIRNA Assays ›
Gene Symbol:
Dync1i2
Gene Name:
dynein cytoplasmic 1 intermediate chain 2
Gene Aliases:
Dnci2, Dncic2
Chromosome Location:
Chr.3: 57817689 - 57868854 on Build Rnor_6.0
UniGene ID:
Rn.11014
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300529
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dync1i2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270624.1 NP_001257553.1 12, 13 1602
NM_001270625.1 NP_001257554.1 11, 12 1542
NM_053880.2 NP_446332.2 13, 14 1620
XM_006234344.2 XP_006234406.1 12, 13 1379
XM_006234346.2 XP_006234408.1 11, 12 1318
GenBank mRNA BC078764.1 - 12, 13 1348 7113840
U39044.1 - 13, 14 1336
U39045.1 - 12, 13 1318
U39046.1 - 11, 12 1258

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001271789, NM_001271787, NM_001271786, NM_001271788, NM_001378, and others.
This siRNA also targets the following transcripts in Mouse: NM_010064, NM_001198876, NM_001198877, NM_001198872, NM_001198878, and others.

Gene Ontology Categories:

Function(s) Process(es)

transport
microtubule-based movement
metabolic process
motor activity
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0