siRNA Details
Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053807.1 | NP_446259.1 | 15 | 1241 | ||
GenBank mRNA | AF274057.1 | - | 15 | 1241 | ||
AF274058.1 | - | 13 | 1106 |

Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053807.1 | NP_446259.1 | 15 | 1241 | ||
GenBank mRNA | AF274057.1 | - | 15 | 1241 | ||
AF274058.1 | - | 13 | 1106 |