This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SORBS2 SIRNA Assays ›
Gene Symbol:
Sorbs2
Gene Name:
sorbin and SH3 domain containing 2
Gene Aliases:
Argbp2
Chromosome Location:
Chr.16: 49626205 - 49820223 on Build Rnor_6.0
UniGene ID:
Rn.24612
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300346
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sorbs2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053770.1 NP_446222.1 6 869
GenBank mRNA AF026505.1 - 6 869

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001205219, NM_172752, BC172140, AK139388, AK083429, and others.

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
cell adhesion
cell migration
protein domain specific binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0