This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTPRE SIRNA Assays ›
Gene Symbol:
Ptpre
Gene Name:
protein tyrosine phosphatase, receptor type, E
Gene Aliases:
PTPepsilon
Chromosome Location:
Chr.1: 207883538 - 207987123 on Build Rnor_6.0
UniGene ID:
Rn.107819
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300324
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptpre
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053767.1 NP_446219.1 12, 13 1306
XM_006230449.2 XP_006230511.1 14, 15 4227
XM_006230450.2 XP_006230512.1 11, 12 1234
XM_006230451.2 XP_006230513.1 12, 13 3538
XM_008759971.1 XP_008758193.1 13, 14 1905
XM_008759972.1 XP_008758194.1 13, 14 2406
GenBank mRNA BC166573.1 - 12, 13 1316 7320594
D78610.1 - 11, 12 1180

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transmembrane receptor protein tyrosine phosphatase signaling pathway
regulation of mast cell activation
peptidyl-tyrosine dephosphorylation
negative regulation of insulin receptor signaling pathway
molecular_function
protein tyrosine phosphatase activity
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0