This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC7A10 SIRNA Assays ›
Gene Symbol:
Slc7a10 Lrp3
Gene Name:
solute carrier family 7 (neutral amino acid transporter light chain, asc system), member 10
low density lipoprotein receptor-related protein 3
Gene Aliases:
Asc-1
-
Chromosome Location:
Chr.1: 91432952 - 91449504 on Build Rnor_6.0
Chromosome Location:
Chr.1: 91447308 - 91462682 on Build Rnor_6.0
UniGene ID:
Rn.128083
Rn.145216
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS300235
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc7a10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053726.2 NP_446178.2 10 1384
XM_006228873.2 XP_006228935.1 9 1312
GenBank mRNA AB126813.1 - 10 1384
BC127467.1 - 10 1355 7313174
Gene Symbol: Lrp3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759197.1 XP_008757419.1 7 5642
XM_008759198.1 XP_008757420.1 8 4633
XM_008759199.1 XP_008757421.1 7 4514
XM_008759200.1 XP_008757422.1 8 4561
XM_008759201.1 XP_008757423.1 7 5521
XM_008759202.1 XP_008757424.1 8 4441

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter apolipoprotein

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
neutral amino acid transport
L-serine transport
D-alanine transport
D-serine transport
endocytosis
biological_process
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
D-serine transmembrane transporter activity
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0