This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDK6 SIRNA Assays ›
Gene Symbol:
Cdk6
Gene Name:
cyclin-dependent kinase 6
Gene Aliases:
-
Chromosome Location:
Chr.4: 27781728 - 27969252 on Build Rnor_6.0
UniGene ID:
Rn.162731
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300178
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdk6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191861.1 NP_001178790.1 6 807
XM_006236019.2 XP_006236081.1 7 1271
XM_006236020.2 XP_006236082.1 7 1205
XM_006236021.1 XP_006236083.1 7 1147
XM_008762679.1 XP_008760901.1 7 1122

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell-matrix adhesion
hematopoietic progenitor cell differentiation
type B pancreatic cell development
protein phosphorylation
Notch signaling pathway
response to virus
regulation of gene expression
positive regulation of gene expression
phosphorylation
hemopoiesis
T cell differentiation in thymus
gliogenesis
cell dedifferentiation
negative regulation of myeloid cell differentiation
regulation of erythrocyte differentiation
negative regulation of osteoblast differentiation
negative regulation of cell cycle
positive regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
hematopoietic stem cell differentiation
negative regulation of cellular senescence
cyclin-dependent protein serine/threonine kinase activity
ATP binding
kinase activity
cyclin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0