This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SC5D SIRNA Assays ›
Gene Symbol:
Sc5d
Gene Name:
sterol-C5-desaturase
Gene Aliases:
Sc5dl
Chromosome Location:
Chr.8: 46528066 - 46537014 on Build Rnor_6.0
UniGene ID:
Rn.17462
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS300100
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sc5d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053642.2 NP_446094.1 6 781
GenBank mRNA AB052846.1 - 5 759
BC081704.1 - 6 781 7131154

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
fatty acid biosynthetic process
cholesterol biosynthetic process via lathosterol
oxidation-reduction process
C-5 sterol desaturase activity
iron ion binding
lathosterol oxidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0