This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM9 SIRNA Assays ›
Gene Symbol:
Trim9
Gene Name:
tripartite motif-containing 9
Gene Aliases:
-
Chromosome Location:
Chr.12: 70244533 - 70347761 on Build GRCm38
UniGene ID:
Mm.184012
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS294672
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trim9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110202.1 NP_001103672.1 1 1223
NM_001110203.1 NP_001103673.1 1 1223
NM_001286386.1 NP_001273315.1 1 1223
NM_001286387.1 NP_001273316.1 1 1223
NM_001286388.1 NP_001273317.1 1 1223
NM_053167.3 NP_444397.2 1 1223
GenBank mRNA AK028280.1 - 1 942
AK035807.1 - 1 941
AK048875.1 - 1 960
AK049907.1 - 1 937
AK129109.1 - 1 928
AK163123.1 - 1 960
BC052034.1 - 1 1223 5684877

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_015163, NM_052978, AK055388, D87458, BC013414, and others.
This siRNA also targets the following transcripts in Rat: NM_130420, XM_008764692, XM_008764691, XM_008764694, XM_008764693, and others.

Gene Ontology Categories:

Function(s) Process(es)

synaptic vesicle exocytosis
protein ubiquitination
negative regulation of SNARE complex assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of calcium ion-dependent exocytosis
SNARE binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
protein homodimerization activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0