This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPANK1 SIRNA Assays ›
Gene Symbol:
Gpank1
Gene Name:
G patch domain and ankyrin repeats 1
Gene Aliases:
Bat-4, Bat4, D17H6S54E, G5
Chromosome Location:
Chr.17: 35121473 - 35124815 on Build GRCm38
UniGene ID:
Mm.7405
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS294463
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpank1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128597.1 NP_001122069.1 3 1168
NM_032460.2 NP_115849.2 2 966
GenBank mRNA BC057902.1 - 3 1127 5027029
BC120637.1 - 2 967 40129560
BC120639.1 - 2 967 40129562

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleic acid binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0