This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other B230219D22RIK SIRNA Assays ›
Gene Symbol:
B230219D22Rik
Gene Name:
RIKEN cDNA B230219D22 gene
Gene Aliases:
AA119856, D530037I19Rik
Chromosome Location:
Chr.13: 55693124 - 55703500 on Build GRCm38
UniGene ID:
Mm.28449
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS294206
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: B230219D22Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181278.2 NP_851795.1 2 430
GenBank mRNA AJ492504.1 - 1 221
AK088693.1 - 2 447
BC051430.1 - 2 430 6530869

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0