This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRR15 SIRNA Assays ›
Gene Symbol:
Prr15
Gene Name:
proline rich 15
Gene Aliases:
E130201N16Rik, G90
Chromosome Location:
Chr.6: 54327012 - 54330200 on Build GRCm38
UniGene ID:
Mm.41984
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS294065
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prr15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030024.2 NP_084300.1 2 703
GenBank mRNA AJ132433.2 - 2 703
AK021401.1 - 2 811
AK136438.1 - 2 883
BC055838.1 - 2 832
BC132416.1 - 1 379 40130693
BC138757.1 - 1 379 8861779

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0