This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700113H08RIK SIRNA Assays ›
Gene Symbol:
1700113H08Rik
Gene Name:
RIKEN cDNA 1700113H08 gene
Gene Aliases:
-
Chromosome Location:
Chr.10: 87040467 - 87230599 on Build GRCm38
UniGene ID:
Mm.78423
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS293699
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700113H08Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029685.1 NP_083961.1 5 703

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header