This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTLL9 SIRNA Assays ›
Gene Symbol:
Ttll9
Gene Name:
tubulin tyrosine ligase-like family, member 9
Gene Aliases:
1700016F23Rik, 4930509O20Rik, AW046572
Chromosome Location:
Chr.2: 152950811 - 153008482 on Build GRCm38
UniGene ID:
Mm.44746
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS292981
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttll9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083618.1 NP_001077087.1 11 910
NM_029064.2 NP_083340.2 11 910
GenBank mRNA AK015740.1 - 11 890
AM690753.1 - 10 854
BC148285.2 - 9 664

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
biological_process
molecular_function
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0