This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAMSTR SIRNA Assays ›
Gene Symbol:
Mamstr
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
2810022D01Rik, 5430432N15Rik, AW743872, Mastr
Chromosome Location:
Chr.7: 45639200 - 45648206 on Build GRCm38
UniGene ID:
Mm.107442
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS292892
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mamstr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172418.2 NP_766006.2 8, 9 1069
GenBank mRNA AK017382.1 - 7, 8 1377
AK036109.1 - 5 1277
AK052976.1 - 2 2312
BC107175.1 - 3, 4 588
BC107176.1 - 3, 4 590
BC119481.1 - 3, 4 588
BC119482.1 - 3, 4 588
BC127227.1 - 3, 4 590
BC127228.1 - 3, 4 590
DQ534901.1 - 8, 9 1053

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of myotube differentiation
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, RNA polymerase II transcription factor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header