This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTLL11 SIRNA Assays ›
Gene Symbol:
Ttll11
Gene Name:
tubulin tyrosine ligase-like family, member 11
Gene Aliases:
4932702F08Rik, 4933424A20Rik, AU043211, D2Ertd624e
Chromosome Location:
Chr.2: 35751226 - 35979624 on Build GRCm38
UniGene ID:
Mm.21591
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS292835
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttll11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029774.2 NP_084050.1 10 2005
GenBank mRNA AK016577.1 - 9 1363
AM690755.1 - 10 2005
BC115858.1 - 7 1355 40057265

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_001071863, BC169049.

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
protein polyglutamylation
microtubule severing
molecular_function
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0