This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TM2D2 SIRNA Assays ›
Gene Symbol:
Tm2d2
Gene Name:
TM2 domain containing 2
Gene Aliases:
2410018G23Rik, Blp1
Chromosome Location:
Chr.8: 25017211 - 25023260 on Build GRCm38
UniGene ID:
Mm.28626
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290920
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tm2d2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027194.3 NP_081470.2 1 182
GenBank mRNA AK010545.1 - 1 114
AK045768.1 - 1 141
AK080854.1 - 1 110
AK083654.1 - 1 103
AK085975.1 - 1 134
AK086015.1 - 1 120
AK086059.1 - 1 136
AK088246.1 - 1 134
AK149407.1 - 1 111
AK160039.1 - 1 119
AK166012.1 - 1 135
BC026789.1 - 1 109 4188168
BY018367.1 - 1 182

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0