This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VKORC1L1 SIRNA Assays ›
Gene Symbol:
Vkorc1l1
Gene Name:
vitamin K epoxide reductase complex, subunit 1-like 1
Gene Aliases:
2310024K08Rik
Chromosome Location:
Chr.5: 129942109 - 129986692 on Build GRCm38
UniGene ID:
Mm.288718
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290821
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vkorc1l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001327.2 NP_001001327.1 2 346
NM_001286382.1 NP_001273311.1 3 318
NM_027121.4 NP_081397.1 3 456
GenBank mRNA AK009497.1 - 2 226
AK078942.1 - 2 289
AY423046.1 - 3 398
BB625262.1 - 2 289
BC085133.1 - 2 346 6733526
BC125315.1 - 3 442 40129830
BC138132.1 - 3 442 8861151
CN701203.1 - 3 296

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_173517, CR627471, AY423045, AK124688, BC027734, and others.
This siRNA also targets the following transcripts in Rat: NM_203338, XM_008766000, XM_008766001, AY423048, AY321342.

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-glutamic acid carboxylation
cellular response to oxidative stress
vitamin K metabolic process
oxidation-reduction process
oxidoreductase activity
vitamin-K-epoxide reductase (warfarin-sensitive) activity
quinone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0