This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMD11 SIRNA Assays ›
Gene Symbol:
Psmd11
Gene Name:
proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
Gene Aliases:
1700089D09Rik, 1810019E17Rik, 2610024G20Rik, 2810055C24Rik, C78232, P44.5, S9
Chromosome Location:
Chr.11: 80428615 - 80473248 on Build GRCm38
UniGene ID:
Mm.260539
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290536
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmd11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178616.3 NP_848731.2 3 280
GenBank mRNA AK007547.1 - 3 249
AK012951.1 - 3 289
AK051738.1 - 3 270
AK076002.1 - 3 268
AK089942.1 - 3 245
AK136049.1 - 3 243
AK140773.1 - 264
AK167559.1 - 3 251
AK214581.1 - 2 134
AK215818.1 - 2 134
BC037590.1 - 3 229
BC055457.1 - 3 212
BC090664.1 - 3 232 30677672
BC090980.1 - 3 247 30283128
BC100595.1 - 3 242 6767671
BC119136.1 - 3 251 8733885
BC119138.1 - 3 251 8733887
BY092952.1 - 3 281

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
proteasome assembly
stem cell differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0