This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC88C SIRNA Assays ›
Gene Symbol:
Ccdc88c
Gene Name:
coiled-coil domain containing 88C
Gene Aliases:
0610010D24Rik, AV000935, AW324073, Daple, mKIAA1509
Chromosome Location:
Chr.12: 100911521 - 101028983 on Build GRCm38
UniGene ID:
Mm.156217
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290181
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccdc88c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026681.4 NP_080957.2 19 3353
GenBank mRNA AK173188.1 - 5 728
AY339881.1 - 19 3354
BC043115.1 - 21 2964 6408630
BC063255.1 - 21 2964 6408630

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
Wnt signaling pathway
stress-activated protein kinase signaling cascade
protein destabilization
protein homooligomerization
protein binding
PDZ domain binding
protein self-association

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0