This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MTURN SIRNA Assays ›
Gene Symbol:
Mturn
Gene Name:
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Gene Aliases:
2410066E13Rik, AI448550, B230212L03Rik
Chromosome Location:
Chr.6: 54681624 - 54703855 on Build GRCm38
UniGene ID:
Mm.233860
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS290143
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price 492.00
Your Price
Online offer:
492.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mturn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289740.1 NP_001276669.1 3 461
NM_001289741.1 NP_001276670.1 3 461
GenBank mRNA AK019120.1 - 3 342
AK019128.1 - 3 340
AK043452.1 - 3 314
BC042507.1 - 3 353 4235651
BC052467.1 - 3 345 6834900

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
neuron development
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0