This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1600012H06RIK SIRNA Assays ›
Gene Symbol:
1600012H06Rik
Gene Name:
RIKEN cDNA 1600012H06 gene
Gene Aliases:
-
Chromosome Location:
Chr.17: 14943184 - 14945939 on Build GRCm38
UniGene ID:
Mm.390949
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS289918
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1600012H06Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083880.1 NP_001077349.1 2 680
NM_001083881.1 NP_001077350.1 2 750
NM_001083882.1 NP_001077351.1 1 932
NM_026451.2 NP_080727.2 2 685
GenBank mRNA AK005427.1 - 2 662
AK017276.1 - 1 862
AK028052.1 - 2 686
AK076083.1 - 2 643
AK076441.1 - 889
AK076476.1 - 2 605
AK083064.1 - 1 873
AK150772.1 - 2 705
AK167490.1 - 2 692
BC027313.1 - 2 664 3710655
BC043303.1 - 1 893
BC043449.1 - 1 893
BC054359.1 - 2 682
BC071250.1 - 2 597

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0