This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1600012H06RIK SIRNA Assays ›
Gene Symbol:
1600012H06Rik
Gene Name:
RIKEN cDNA 1600012H06 gene
Gene Aliases:
-
Chromosome Location:
Chr.17: 14943184 - 14945939 on Build GRCm38
UniGene ID:
Mm.390949
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS289916
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1600012H06Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083880.1 NP_001077349.1 2 308
NM_001083881.1 NP_001077350.1 2 378
NM_001083882.1 NP_001077351.1 1 560
NM_026451.2 NP_080727.2 2 313
GenBank mRNA AK005427.1 - 2 290
AK017276.1 - 1 490
AK018298.1 - 2 236
AK028052.1 - 2 314
AK076083.1 - 2 270
AK076441.1 - 517
AK076476.1 - 2 233
AK083064.1 - 1 501
AK150772.1 - 2 333
AK167490.1 - 2 320
BB610149.1 - 2 314
BC027313.1 - 2 292 3710655
BC043303.1 - 1 521
BC043449.1 - 1 521
BC054359.1 - 2 310
BC071250.1 - 2 225

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001103356, XM_006227969, XM_006227970, XM_008758746, BC079175.

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0