This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other COA6 SIRNA Assays ›
Gene Symbol:
Coa6
Gene Name:
cytochrome c oxidase assembly factor 6
Gene Aliases:
1810063B05Rik, AI447995
Chromosome Location:
Chr.8: 126422501 - 126425435 on Build GRCm38
UniGene ID:
Mm.31946
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS289901
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Coa6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174987.4 NP_778152.1 1 249
GenBank mRNA AK008041.1 - 1 184
AK009404.1 - 1 223
BC024399.1 - 1 179 5026081
BC038634.1 - 1 235 1513986
BC046907.1 - 2 140 6394880
BU562971.1 - 1 252
CO039015.1 - 2 528

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
cytochrome-c oxidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0