siRNA Details
Gene Symbol: 4930528A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014838.1 | - | 1 | 266 | ||
AK015922.1 | - | 1 | 186 | |||
AK039347.1 | - | 1 | 248 | |||
BC031206.1 | - | 1 | 233 |

Gene Symbol: 4930528A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014838.1 | - | 1 | 266 | ||
AK015922.1 | - | 1 | 186 | |||
AK039347.1 | - | 1 | 248 | |||
BC031206.1 | - | 1 | 233 |