This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXO32 SIRNA Assays ›
Gene Symbol:
Fbxo32
Gene Name:
F-box protein 32
Gene Aliases:
4833442G10Rik, AI430017, ATROGIN1, Gm20361, MAFbx
Chromosome Location:
Chr.15: 58175879 - 58214892 on Build GRCm38
UniGene ID:
Mm.25683
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID MSS289756
Size 20 nmol
Availability Made To Order
Catalog # 1320001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxo32
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026346.3 NP_080622.1 9 1327
GenBank mRNA AF441120.1 - 9 1373
AK014612.1 - 9 1280
AK014793.1 - 9 1309
AK190642.1 - 3 260
BC027211.1 - 9 1261 3981597

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to electrical stimulus involved in regulation of muscle adaptation
muscle atrophy
response to denervation involved in regulation of muscle adaptation
cellular response to dexamethasone stimulus
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0